Learn More
Praseodymium(III) nitrate hydrate, 99.9% (REO), Thermo Scientific Chemicals
36.68 € - 1060.28 €
Chemical Identifiers
CAS | 15878-77-0 |
---|---|
Molecular Formula | H12N3O15Pr |
Molecular Weight (g/mol) | 435.01 |
MDL Number | MFCD00011175 |
InChI Key | LXXCECZPOWZKLC-UHFFFAOYSA-N |
Synonym | praseodymium iii nitrate hexahydrate, praseodymium iii nitrate, hexahydrate 1:3:6, praseodymium nitrate hexahydrate, nitric acid, praseodymium 3+ salt, hexahydrate, praseodymium 3+ hexahydrate trinitrate, ksc529q4r, praseodymium trinitrate hexahydrate, praseodymium nitrate hydrate 1:3:6, praseodymium nitrate,, praseodymium 3+ trinitrate hexahydrateShow More |
PubChem CID | 204170 |
SMILES | O.O.O.O.O.O.[Pr+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11317647
|
Thermo Scientific Alfa Aesar
012909.09 |
10 g |
36.68 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11397637
|
Thermo Scientific Alfa Aesar
012909.18 |
50 g |
102.51 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11307647
|
Thermo Scientific Alfa Aesar
012909.30 |
250 g |
375.87 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11387637
|
Thermo Scientific Alfa Aesar
012909.A1 |
1 kg |
1060.28 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Praseodymium(III) nitrate hydrate is used in fluorescent display tubes and phosphors. It is also used in the ultrasonic synthesis of phase-pure praseodymium molybdate. It plays a major role in the solvothermal process for formation of doped lanthanide oxysulfide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
15878-77-0 | |
435.01 | |
LXXCECZPOWZKLC-UHFFFAOYSA-N | |
204170 |
H12N3O15Pr | |
MFCD00011175 | |
praseodymium iii nitrate hexahydrate, praseodymium iii nitrate, hexahydrate 1:3:6, praseodymium nitrate hexahydrate, nitric acid, praseodymium 3+ salt, hexahydrate, praseodymium 3+ hexahydrate trinitrate, ksc529q4r, praseodymium trinitrate hexahydrate, praseodymium nitrate hydrate 1:3:6, praseodymium nitrate,, praseodymium 3+ trinitrate hexahydrateShow More | |
O.O.O.O.O.O.[Pr+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Specifications
15878-77-0 | |
Crystalline | |
H12N3O15Pr | |
praseodymium iii nitrate hexahydrate, praseodymium iii nitrate, hexahydrate 1:3:6, praseodymium nitrate hexahydrate, nitric acid, praseodymium 3+ salt, hexahydrate, praseodymium 3+ hexahydrate trinitrate, ksc529q4r, praseodymium trinitrate hexahydrate, praseodymium nitrate hydrate 1:3:6, praseodymium nitrate,, praseodymium 3+ trinitrate hexahydrate | |
LXXCECZPOWZKLC-UHFFFAOYSA-N | |
praseodymium(3+) hexahydrate trinitrate | |
204170 | |
Odorless | |
Green crystals |
Green | |
99.9% | |
MFCD00011175 | |
Soluble in water,anhydrous amine,alcohol,ether,polar solvents and acetonitrile. | |
O.O.O.O.O.O.[Pr+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
435.01 | |
326.92 (Anhydrous) | |
Hygroscopic | |
Praseodymium(III) nitrate hydrate, Rare Earth Oxides Content |
Safety and Handling
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.
P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H272-H315-H319-H335
missing translation for 'dotInformation' : DOT Class: 5.1, Packaging Group: III
missing translation for 'einecsNumber' : 233-796-5
missing translation for 'rtecsNumber' : TU1400000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
- Praseodymium(III) nitrate hydrate
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Oxidising solid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H272-May intensify fire; oxidiser.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P220-Keep/Store away from clothing/combustible materials.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Praseodymium(III) nitrate hexahydrate
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.