Learn More
Thermo Scientific™ ProSpecT™ Giardia EZ Microplate Assay
Rapidly detect Giardia specific antigen from human fecal samples.
Brand: Thermo Scientific™ R2458596
Description
Rapidly detect Giardia specific antigen in fresh or preserved human fecal specimens using Thermo Scientific™ ProSpecT™ Giardia EZ Microplate Assay.
Manage your resources, increase productivity and improve workflow efficiency with this direct, qualitative enzyme immmunoassay for the detection of Giardia. It combines excellent sensitivity/specificity, a common procedure with no indeterminate range, plus flexible specimen transport options.- Excellent clinical performance
- Cary Blair transport option which saves a dilution step
- Results within 100 minutes
- Room temperature incubation
- True colorless negatives
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

Specifications
2°C to 8°C | |
Giardia | |
Giardia Microplate Assay | |
Transfer pipettes, microplate strip holder and cover, microplate (8 wells/strip), enzyme conjugate, positive control, negative control, sample diluent, wash buffer, color substrate, stop solution and procedure card |
ProSpecT Giardia EZ Microplate Assay | |
Fecal | |
Enzyme Immunoassay Detection | |
96 Wells/Kit |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.