missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ PSC Cryopreservation Kit
PSC Cryopreservation Kit
Brand: Gibco™ A2644601
This item is not returnable.
View return policy
Description
The PSC Cryopreservation Kit contains xeno-free PSC Cryopreservation Medium, which is a ready-to-use solution for the cryopreservation of early passage pluripotent stem cells (PSCs), and RevitaCell™ Supplement (100X), a chemically defined recovery supplement for use in the post-thaw culture media. When used in combination, these reagents help minimize loss of cell viability, maximize post-thaw recovery, and minimize unwanted differentiation of pluripotent stem cells. This kit can also be used to cryopreserve and recover peripheral blood mononuclear cells (PBMCs) to improve post-thaw cell viability and recovery.Reliable
Xeno-free, chemically defined formulations help ensure the proliferation capacity and long-term culture of PSCs and PBMCs in a stable state, thus helping to increase their usefulness in research and clinical research applications.
Easy to use
The PSC Cryopreservation Medium is provided as a ready-to-use, complete solution containing 10% DMSO for efficient cell addition. Separate additions of enhancing reagents are not required, helping save valuable preparation time.
Versatile
The kit is compatible with cells cultured in a variety of media systems including Essential 8™ medium, StemPro™ hESC SFM, and other commercially available formulations. Also, the kit is compatible with clump and single cell passaged PSCs.
cGMP manufacturing and quality system
The PSC Cryopreservation Kit is manufactured at a cGMP-compliant facility. The facility is also registered with the FDA as a medical device manufacturer and certified to ISO 13485 standards.

Specifications
• 50 mL Cryopreservation Media • 5 mL RevitaCell Supplement (100X) Store at -5 to -20°C and protect from light. |
|
Stem Cell (Human, iPS - Induced Pluripotent Stem, Embryonic) | |
Serum-free | |
cGMP for medical devices, 21 CFR Part 820 and ISO 13485 | |
50 mL |
Xeno-free for Cryomedium, Animal Origin-free for RevitaCell | |
PSC Cryopreservation Kit | |
DMSO (10%) | |
Gibco™ | |
Dry Ice |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 10
User Guide: PSC Cryopreservation Kit
Version: MAN0013466 Rev A.0 (05 February 2015)
File size: 133.4 kB
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Gibco™ PSC Cryopreservation Kit > 50mL
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction