missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ StemFlex™ Medium

Supports the robust expansion of feeder-free pluripotent stem cells (PSCs). Optimized to support novel applications, including single-cell passaging, gene editing, and reprogramming. StemFlex Medium’s formulation offers the convenience of a flexible feeding schedule (including weekend-free options).

Brand:  Gibco™ A3349401

Product Code. 15627578

  • 454.00 â‚¬ / 500mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Flexibility: StemFlex Medium allows for unprecedented 3-way usage flexibility for PSC culture conditions: the ability to select an optimal feeding schedule, including a weekend-free option; a choice of matrices, depending upon the needs of the workflow; and the option to select the passaging reagent for the level of cell dissociation required.
  • Exceptional value and performance: StemFlex Medium is provided in a convenient two-component kit (450mLbasal medium and 50mL supplement), and when used with Geltrex™ LDEV-Free, hESC-Qualified, Reduced Growth Factor Basement Membrane Matrix, provides a cost-effective, robust system for superior feeder-free culture of human PSCs. StemFlex Medium’s ability to enable weekend-free culture of PSCs provides unmatched value among the robust feeder-fee PSC media on the market today
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Embryonic Stem Cells, , Induced Pluripotent Stem Cells
Phenol Red
StemFlex™
Supplement ships on dry ice, basal can ship at RT
Feeder-free Stem Cell Culture (Human, iPS - Induced Pluripotent Stem, Embryonic)
Stem Cell Media
cGMP for medical devices, 21 CFR Part 820 and ISO 13485
500 mL
• 450 mL Basal Medium, store at 2–8°C and protect from light
• 50 mL Supplement, store at -5 to -20°C and protect from light
Product Suggestions

Product Suggestions

Videos

Videos

  • Gibco StemFlex medium benefits
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
     
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected

      Gibco StemFlex medium benefits

    SDS
    Documents Certificates
    Special Offers

    Special Offers

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title
    Gibco™ StemFlex™ Medium > StemFlex™ Medium

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

    For Research Use Only. Not for use in diagnostic procedures.