missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTFE Beaker Lids
Click to view available options
For Use With (Equipment):
1000mL beaker
100mL beaker
10mL beaker
2000mL beaker
250mL beaker
25mL beaker
500mL beaker
50mL beaker
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Available in a range of diameters

Specifications
Specifications
Diameter (Metric) Outer | 100mm |
Material | PTFE |
For Use With (Equipment) | 1000mL beaker |
Height (Metric) | 155mm |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 9
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PTFE Beaker Lids >
Spot an opportunity for improvement?Share a Content Correction