missing translation for 'onlineSavingsMsg'
Learn More

Masterflex™ L/S™ Standard Pump Head for High-Performance Precision Tubing L/S™ 35, Polycarbonate Housing, SS Rotor

Ideal for fluid transfer applications requiring high accuracy and repeatability

Brand:  Masterflex™ 07035-21

Product Code. 11701209

  • 487.00 € / Each
Estimated Shipment: 20-06-2025
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Standard heads are precision molded to accommodate a single size of tubing
  • The pump head is compact and simple in design with same side tubing entry/exit—perfect for space-limited applications
  • Standard heads are compact and easy to integrate into your OEM applications
  • Stack up to four heads depending on drive recommendations
  • The rotor is constructed of stainless steel (SS) for enhanced durability
  • Suitable for use with L/S™ 35 High-Performance Precision pump tubing
Specifications

Specifications

Standard pump head for L/S 35 tubing, PC housing/SS rotor.
Polycarbonate
3
3 in.
2.5 in.
3 in.
Stainless Steel
Volume Per Revolution With L/S 35 Tubing (mL): 3.8 mL
0.02 to 600 rpm
2300 mL/min.
1
Fixed
77 mm
64 mm
77 mm
L/S 35
Stainless Steel
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Masterflex™ L/S™ Standard Pump Head for High-Performance Precision Tubing L/S™ 35, Polycarbonate Housing, SS Rotor >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.