missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrex™ Aspirator Bottle with Tubing Side Arm
Store and discharge a variety of liquids with this robust aspirator bottle. Pyrex™ Aspirator Bottles with Tubing Side Arm are useful as a delivery or storage container for a wide range of solutions. These bottles feature sturdy ring necks with a heavy duty rims.
49.77 € - 636.00 €
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
10355140
|
Pyrex
1531/10D |
1000 mL |
420.16 €
Pack of 5 |
Estimated Shipment: 03-06-2025
Log in to see stock. |
||||||
11317884
|
Pyrex
1531/10D |
1000 mL | N/A | N/A | N/A | |||||
11307884
|
Pyrex
1531/08D |
500 mL | N/A | N/A | N/A | |||||
13471829
|
Pyrex
1531/08D |
500 mL |
636.00 €
Pack of 10 |
Estimated Shipment: 30-05-2025
Log in to see stock. |
||||||
10599530
|
Pyrex
1531/06D |
250 mL | N/A | N/A | N/A | |||||
10191250
|
Pyrex
1531/12D |
2000 mL |
554.32 €
Pack of 5 |
Estimated Shipment: 03-06-2025
Log in to see stock. |
||||||
Description
- Tubular side arm for attachment of 9mm bore flexible tubing
- Unground neck to accept rubber stoppers
- Withstands repeated sterilization (wet or dry)
Specifications
Borosilicate Glass | |
Aspirator Bottle |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Pyrex™ Aspirator Bottle with Tubing Side Arm