missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrex™ Equalizing Glass Graduated Funnel with PTFE Key
For use when liquids have to be added to systems under vacuum.
Brand: Pyrex 443533
This item is not returnable.
View return policy
Description
- Complies with ISO 4800, Type 4
- With interchangeable PTFE key for grease free performance
- Double graduated scale showing volume of liquid contained/delivered
- Stem inside cone for easy fitting to Quickfit™ assemblies
- Graduations in white enamel
- Supplied with hollow glass stopper

Specifications
50 mL | |
Borosilicate Glass | |
Yes | |
Pressure-equalizing Funnel |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Pyrex™ Equalizing Glass Graduated Funnel with PTFE Key > Capacity: 50mL
Spot an opportunity for improvement?Share a Content Correction