missing translation for 'onlineSavingsMsg'
Learn More

PYREX™ Pear-Shaped Glass Separating Funnel with PTFE Key

Manufactured from Pyrex™ glass for corrosion-free performance. Pyrex™ Pear-Shaped Glass Separating Funnel with Glass Key complies with ISO 4800 Type 2 Standards. Includes glass stopper and interchangeable PTFE key.

Brand:  Pyrex 2180/04M

98.71 EUR valid until 2025-06-13
Use promo code "24123" to get your promotional price.


 View more versions of this product

Product Code. 11353684

  • 112.00 € / Each
Estimated Shipment: 10-07-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Manufactured from Pyrex™ glass for corrosion-free performance. Pyrex™ Pear-Shaped Glass Separating Funnel with Glass Key complies with ISO 4800 Type 2 Standards. Includes glass stopper and interchangeable PTFE key.

  • Complies with ISO 4800 Type 2
  • Manufactured from Pyrex™ glass for corrosion-free performance
  • With interchangeable PTFE key
  • Supplied with glass stopper
TRUSTED_SUSTAINABILITY
Specifications

Specifications

100 mL
ISO 4800
1.5mm; Glass
19/26
Borosilicate Glass
1PK
Glass
Separatory Funnel
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
PYREX™ Pear-Shaped Glass Separating Funnel with PTFE Key > Capacity: 100mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.