missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Qdot™ 655 Streptavidin Conjugate

Qdot™ 655 Streptavidin Conjugate

Brand:  Molecular Probes™ Q10121MP

679.44 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.


 View more versions of this product

Product Code. 10591855

  • 894.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

1 μM
200 μL
Room Temperature
Qdot™
Qdot 655
Store in refrigerator (2–8°C).
Liquid
Streptavidin Conjugate (fluorescent)
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2831248 Certificate TypeCertificate of Analysis Date14/03/2024 Catalog NumberQ10121MP
Lot Number2663880 Certificate TypeCertificate of Analysis Date18/09/2023 Catalog NumberQ10121MP
Lot Number2486542 Certificate TypeCertificate of Analysis Date09/06/2022 Catalog NumberQ10121MP
Lot Number2354407 Certificate TypeCertificate of Analysis Date15/06/2021 Catalog NumberQ10121MP
Lot Number2305180 Certificate TypeCertificate of Analysis Date12/01/2021 Catalog NumberQ10121MP
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Molecular Probes™ Qdot™ 655 Streptavidin Conjugate > 200μL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.