Learn More
PURUS™ Critical Print Cleanroom Notebooks
Critical Print Notebooks are ideal for writing and data recording in cleanroom environments.
Brand:  PURUS™ PNBCPA62240 WG
 View more versions of this product
Product Code. 15783609
  • 365.00 € / Pack of 40
Estimated Shipment: 17-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
For writing in cleanroom environments
  • Cleanroom processed and packaged
  • Suitable for use in ISO 4 to 6 (Class 10 to Class 1000) cleanrooms
  • Proprietary Binding Technology without organic fillers
  • Recyclable
  • College Rule
  • Amide and Silicone Free
Specifications
Specifications
Polyethylene
White
148 x 105 mm
Autoclavable
For writing in cleanroom environments
40/Cs.
80 gsm
Grid
Poly
105 mm
Suitable for use in ISO 4 to 6 (Class 10 to Class 1000)
Cleanroom Notebook
A6
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
PURUS™ Critical Print Cleanroom Notebooks > White, A6, Grid