missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ QualiSampler™ Set

The handy set, which comes complete with carrying case, combines the LiquiSampler and the ViscoSampler with practical accessories.Models in PP or PTFE/ FEP.

Brand:  Buerkle™ 5330-1001

 View more versions of this product

Product Code. 10329630

  • 1250.00 € / Each
Estimated Shipment: 09-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

QualiSampler set consisting of:

  • LiquiSampler, (Immersion depth: 100 cm, Capacity: 250mL)
  • ViscoSampler, 100 cm (Immersion depth: 100 cm, Capacity: 300mL)
  • Case, five bottles, cleaning brush
Specifications

Specifications

LiquiSampler
PTFE, FEP
100 cm PTFE/FEP LiquiSampler, 100 cm PTFE/FEP viscosampler, case, five bottles, cleaning brush
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Buerkle™ QualiSampler™ Set > PTFE/FEP

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.