Learn More
Pyrex™ Quickfit™ Adjustable Wash Bottle Head
Used in conjunction with a Drechsel bottle for gas-washing purposes. Pyrex™ Quickfit™ Adjustable Wash Bottle Head allows use with either a 125mL or 250mL MF29 series wash bottle.
Brand: Pyrex™ MF27/3/13
61.29 EUR valid until 2025-07-13
Use promo code "24135" to get your promotional price.
Description
- With screwthread joint which allows height adjustment of the dip tube and hence permits use with either 125mL or 250mL Dreschel bottles
- To fit MF29 series wash bottles

Specifications
Sidearm O.D.: 7mm, Key: 3, 10 | |
1/Ea. | |
24/29 | |
Wash bottle head |
125ml or 250ml Drechsel bottles | |
Thread size: 13 | |
Borosilicate Glass |
Product Suggestions
Customers who viewed this item also viewed.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.