missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Quickfit™ Two-Neck Round-Bottom Glass Flask
Manufactured from borosilicate glass. Quickfit Jointed Ware™ Quickfit™ Two-Neck Round-Bottom Glass Flask has high resistance to thermal shock and chemical attack.
Brand: Quickfit FR2L/5S/2A
545.30 EUR valid until 2025-06-13
Use promo code "24123" to get your promotional price.
This item is not returnable.
View return policy
Description
Manufactured from borosilicate glass. Quickfit Jointed Ware™ Quickfit™ Two-Neck Round-Bottom Glass Flask has high resistance to thermal shock and chemical attack.
- Fitted with two Quickfit™ ground glass sockets - One vertical centre neck - One angled side neck
- Side neck useful for introducing gas inlet tubes and thermometers etc. , into the flask
- Manufactured from borosilicate glass for highest resistance to thermal shock and chemical attack

Specifications
2000 mL | |
225 mm | |
34/35 (Centre), 19/26 (side) |
Borosilicate Glass | |
2 | |
Round |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product information
File size: 2.8 MB
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Quickfit™ Two-Neck Round-Bottom Glass Flask > Capacity: 2000mL; Socket: 34/35 (Centre), 19/26 (side)
Spot an opportunity for improvement?Share a Content Correction