Learn More
Quickfit™ Two-Neck Round-Bottom Glass Flask
Manufactured from borosilicate glass. Quickfit Jointed Ware™ Quickfit™ Two-Neck Round-Bottom Glass Flask has high resistance to thermal shock and chemical attack.
53.85 € - 574.00 €
Product Code | Brand | Capacity (Metric) | Socket Joint | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Socket Joint | Price | Quantity & Availability | |||||
12030643
|
Quickfit
FR100/3S/2A |
100 mL | 24/29 (Centre), 19/26 (side) |
71.30 €
Each |
Estimated Shipment: 06-06-2025
Log in to see stock. |
|||||
12478966
|
Quickfit
FR100/3S/1A |
100 mL | 24/29 (Centre), 14/23 (side) |
74.20 €
Each |
In Stock
Log in to see stock. ![]() |
|||||
12418996
|
Quickfit
FR250/3S/1A |
250 mL | 24/29 (Centre), 14/23 (side) |
53.85 €
Each |
Estimated Shipment: 20-06-2025
Log in to see stock. |
|||||
12070653
|
Quickfit
FR250/3S/2A |
250 mL | 24/29 (Centre), 19/26 (side) |
71.10 €
Each |
In Stock
Log in to see stock. ![]() |
|||||
12438986
|
Quickfit
FR25/1S/1A |
25 mL | 14/23 (Centre), 14/23 (side) |
54.85 €
Each |
Estimated Shipment: 18-07-2025
Log in to see stock. |
|||||
12050673
|
Quickfit
FR500/3S/2A |
500 mL | 24/29 (Centre), 19/26 (side) |
73.55 €
Each |
Estimated Shipment: 06-06-2025
Log in to see stock. |
|||||
12403631
|
Quickfit
FR50/1S/1A |
50 mL | 14/23 (Centre), 14/23 (side) |
78.05 €
Each |
Estimated Shipment: 18-07-2025
Log in to see stock. |
|||||
15547880
|
Quickfit
FR2L/5S/2A |
2000 mL | 34/35 (Centre), 19/26 (side) |
574.00 €
Pack of 5 |
Estimated Shipment: 21-08-2025
Log in to see stock. |
|||||
15517860
|
Quickfit
FR1L/3S/1A |
1000 mL | 24/29 (Centre), 14/23 (side) |
363.00 €
Pack of 5 |
Estimated Shipment: 30-07-2025
Log in to see stock. |
|||||
15537860
|
Quickfit
FR1L/3S/2A |
1000 mL | 24/29 (Centre), 19/26 (side) |
363.00 €
Pack of 5 |
Estimated Shipment: 06-06-2025
Log in to see stock. |
|||||
15597890
|
Quickfit
FR500/3S/1A |
500 mL | 24/29 (Centre), 14/23 (side) |
292.00 €
Pack of 5 |
Estimated Shipment: 06-06-2025
Log in to see stock. |
|||||
Description
Manufactured from borosilicate glass. Quickfit Jointed Ware™ Quickfit™ Two-Neck Round-Bottom Glass Flask has high resistance to thermal shock and chemical attack.
- Fitted with two Quickfit™ ground glass sockets - One vertical centre neck - One angled side neck
- Side neck useful for introducing gas inlet tubes and thermometers etc. , into the flask
- Manufactured from borosilicate glass for highest resistance to thermal shock and chemical attack
Specifications
Borosilicate Glass | |
2 | |
Round |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Quickfit™ Two-Neck Round-Bottom Glass Flask