Learn More
Quickfit™ Funnel with Ground Glass Cone
Manufactured from chemically resistant Pyrex™ Borosilicate Glass. Quickfit™ Funnel with Ground Glass Cone is designed for transferring powders into Quickfit™ flasks and vessels.
Brand:  Quickfit CF5/2
48.92 EUR valid until 2025-06-13
Use promo code "24123" to get your promotional price.
 View more versions of this product
Product Code. 12004223
  • 57.00 € / Each
Estimated Shipment: 09-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Manufactured from chemically resistant Pyrex™ Borosilicate Glass. Quickfit™ Funnel with Ground Glass Cone is designed for transferring powders into Quickfit™ flasks and vessels.
  • Designed for transferring powders into Quickfit™ flasks and vessels
  • With Quickfit™ ground glass cone
  • Manufactured from chemically resistant Pyrex™ Borosilicate Glass
TRUSTED_SUSTAINABILITY
Specifications
Specifications
105mm
19/26
Powder Funnel
Borosilicate Glass
Quickfit™ Ground Glass Cone
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Quickfit™ Funnel with Ground Glass Cone > Cone Size: 19/26