Learn More
Contec™ Quiltec™ I Wipes
Clean interior surfaces, tools, or products. Quiltec™ I Wipes offer an extremely high sorptive capacity. These durable polyester wipes are ideal for cleanroom applications.
Brand:  Contec™ LWQS0001
 View more versions of this product
Product Code. 16675524
  • 1150.00 € / Pack of 1000
Estimated Shipment: 09-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Clean interior surfaces, tools, or products. Quiltec™ I Wipes offer an extremely high sorptive capacity. These durable polyester wipes are ideal for cleanroom applications.
  • These sealed-edge wipes offer an extremely high sorptive capacity. Two layers of quilted polyester fabric are laser-cut and sealed together to create pockets where additional fluid can be trapped.
  • Two-ply quilted polyester construction creates durable wiper with exceptional absorbency—absorbs more than twice the capacity of a single-layer wiper
  • Extremely low in both particles and fibers—ideal for interior surface cleaning of process tools, wiping down products, or general-purpose cleaning
Order Info
Laundered and packaged in an ISO Class 4 cleanroom.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cleanroom Wipe
Not Autoclavable
Single-use
Cleanroom
2-ply
Polyester
ISO Class 3 and 4
23 x 23 cm (9 x 9 in.)
25/Bag, 40 Bags/Cs.
Sterile
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Contec™ Quiltec™ I Wipes > Dimensions (L x W): 23 x 23 cm (9 x 9 in.), Quantity: 25/Bag, 40 Bags/Cs., Sterile: Yes