Learn More
Welch Ilmvac™ Vacuum Pump Oil
One and two-stage rotary vane pump. Fisherbrand™ Vacuum Pump Oil designed for use with the range of Fisherbrand two-stage rotary vane pumps and with most other one and two-stage rotary vane pumps.
Brand:  Welch Ilmvac™ 800120-01
 View more versions of this product
Product Code. 10777544
  • 137.00 € / 5 L
Explore more special offers
Description
Description
Generic mineral oil for use in most one and two stage rotary vane pumps.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Vacuum Pump Oil
10-5 mabr vapour pressure
5 L
For General Use in Rotary Vane Pumps
118 cSt viscosity at 40°C
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Welch Ilmvac™ Vacuum Pump Oil > Volume: 5L