Learn More
Thermo Scientific™ Racks and Inserts for Refrigerated and Heated Bath Circulators
These stainless-steel racks and inserts support a range of tubes and are designed for use with Thermo Scientific™ Refrigerated and Heated Bath Circulators.
Brand:  Thermo Scientific™ 1600082
 View more versions of this product
Product Code. 11907284
  • 187.94 € / Pack of 2
Estimated Shipment: 01-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
These stainless-steel racks and inserts support a range of tubes and are designed for use with Thermo Scientific™ Refrigerated and Heated Bath Circulators.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Rack Insert; Includes top and bottom panel
Rack Insert
Stainless Steel
Bath Types A25B, A40, S21, S30
13 25 mm Tubes
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Racks and Inserts for Refrigerated and Heated Bath Circulators >