Learn More
Thermo Scientific Chemicals RBC lysis buffer for human
For lysis of human red blood cells
Brand: Thermo Scientific Chemicals J62990.AP
101.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
For lysis of human red blood cells and can be used to lyse reticulocytes, erythrocytes, and mature RBCs.
RBC Lysis Buffer for human is used in the isolation of deoxyribonucleic acid (DNA) and ribonucleic acid (RNA).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
500mL | |
Cell Lysis Reagent | |
2° to 8°C |
Liquid | |
For lysis of human red blood cells. |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
M11L552 | Certificate of Analysis | 21/01/2025 | J62990.AK,J62990.AP |
Z14K531 | Certificate of Analysis | 02/12/2024 | J62990.AK,J62990.AP |
Y18K534 | Certificate of Analysis | 20/11/2024 | J62990.AK,J62990.AP |
Q12K502 | Certificate of Analysis | 11/06/2024 | J62990.AK,J62990.AP |
M16K506 | Certificate of Analysis | 14/02/2024 | J62990.AK,J62990.AP |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only