Learn More
Julabo™ Recirculating Cooling Baths with Water Compressor
Used for routine cooling applications both within laboratory and wider industrial workplace. Julabo™ Recirculating Cooling Baths with Water Compressor has large, bright LED display.
10645.00 € - 28070.00 €
Product Code | Brand | Capacity (Metric) | Flow Rate | Cooling Capacity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Flow Rate | Cooling Capacity | Price | Quantity & Availability | |||
11944818
|
Julabo™
9671017 |
12 to 17 L | 23 L/min. | 0.4kW at -20°C, 0.85kW at -10°C, 1.1kW at 0°C, 1.5kW at 10°C, 1.7kW at 20°C (Med |
10645.00 €
Each |
Estimated Shipment: 29-01-2025
Log in to see stock. |
|||
11924838
|
Julabo™
950002603P3H0 |
21 to 33 L | 33 L/min. | 0.9kW at -10°C, 1.5kW at 0°C, 2.5kW at 20°C (Medium Ethanol) |
25250.00 €
Each |
Estimated Shipment: 29-01-2025
Log in to see stock. |
|||
11994828
|
Julabo™
9676025 |
24 to 30 L | 60 L/min. | 0.3kW at -10°C, 1kW at 0°C, 1.9kW at 10°C, 2.5kW at 20°C (Medium Ethanol) |
16060.00 €
Each |
Estimated Shipment: 29-01-2025
Log in to see stock. |
|||
11984828
|
Julabo™
9673040 |
24 to 30 L | 40 L/min. | 0.45kW at -20°C, 1.3kW at -10°C, 2.2kW at 0°C, 3kW at 10°C, 4.3kW at 20°C (Mediu |
17010.00 €
Each |
Estimated Shipment: 29-01-2025
Log in to see stock. |
|||
11954828
|
Julabo™
9673025 |
24 to 30 L | 40 L/min. | 0.4kW at -20°C, 1kW at -10°C, 1.7kW at 0°C, 2.5kW at 10°C, 2.7kW at 20°C (Medium |
14040.00 €
Each |
Estimated Shipment: 29-01-2025
Log in to see stock. |
|||
11984838
|
Julabo™
9676070 |
39 to 47 L | 60 L/min. | 1.3kW at -20°C, 3.1kW at -10°C, 5.5kW at 0°C, 7kW at 10°C, 7.4kW at 20°C (Medium |
24080.00 €
Each |
Estimated Shipment: 29-01-2025
Log in to see stock. |
|||
11944838
|
Julabo™
950005107P3H0 |
43 to 60 L | 33 L/min. | 1.2kW at -10°C, 2.5kW at 0°C, 5kW at 20°C (Medium Ethanol) |
28035.00 €
Each |
Estimated Shipment: 29-01-2025
Log in to see stock. |
|||
15204373
|
Julabo™
9601063 |
6 to 8 L | 22 L/min. | 0.1kW at -10°C, 0.3kW at 5°C, 0.37kW at 10°C, 0.5kW at 20°C (Medium Ethanol) |
11540.00 €
Each |
Estimated Shipment: 29-01-2025
Log in to see stock. |
|||
11934848
|
Julabo™
9676110 |
39 to 47 L | 60 L/min. | 2.7kW at -20°C, 4.8kW at -10°C, 7.3kW at 0°C, 9kW at 10°C, 11.5kW at 20°C (Mediu |
28070.00 €
Each |
Estimated Shipment: 29-01-2025
Log in to see stock. |
|||
15294363
|
Julabo™
9601060 |
6 to 8 L | 20 L/min. | 0.21kW at -10°C, 0.4kW at 5°C, 0.47kW at 10°C, 0.6kW at 20°C (Medium Ethanol) |
10825.00 €
Each |
Estimated Shipment: 29-01-2025
Log in to see stock. |
|||
11964828
|
Julabo™
9601256 |
8 to 11 L | 28 L/min. | 0.25kW at -20°C, 0.8kW at -10°C, 1.8kW at 0°C, 2kW at 10°C, 2.5kW at 20°C (Mediu |
16000.00 €
Each |
Estimated Shipment: 29-01-2025
Log in to see stock. |
|||
Description
- Ergonomic design and easy operation
- Splashproof keypad with integrated mains switch
- Reliable microprocessor PID temperature control
- Filling level indicator
- Powerful immersion pumps, suitable for continuous operation
- Permissible temperature in return line +80°C
- Easy filling from the top with hinged protective lid
- Low liquid level protection with optical and audible alarm signal
- Integrated stainless steel bath tanks
- Removable venting grid for cleaning of the condenser
- Front drain
- No side vents
- RS232 interface for PC-connection
- IP class according to IEC 60529: 21
- Pressure indicator
- Adjustable bypass for pump pressure
- Water cooled
- Alarm output, potential-free change-over contact (max. 30 VA)