missing translation for 'onlineSavingsMsg'
Learn More

Reichert™ Industrial Fluid Testing Refractometers

Designed specifically for checking ratio or concentration of water-soluble industrial fluids

Brand:  Reichert™ 13754000

Product Code. 10491825

  • 272.00 € / Each
Estimated Shipment: 26-05-2025
to see stock.
Explore more special offers
Add to basket

Includes:
  • Durable plastic carrying case
  • Sample application pipets
  • Calibration adjustment wrench
This item is not returnable. View return policy

Description

Description

Portable testers are ideal for applications including metalworking coolants and lubricants, heat-treating fluids, hydraulic fluids, firefighting foam and plating baths.

  • Automatic temperature compensation provides accurate readings without correction tables or temperature measurement
  • Temperature-compensation range, 15° to 38°C (59° to 100°F)
  • Large, easy-to-read scales and depth of field
  • Portable: no batteries or power cord required
  • Rugged, chemical-resistant housing

IFT10HP Model

  • High Precision optical instrument with improved 0-10 scale 0.1 scale increments for testing low concentration synthetic cutting and grinding fluids and lubricants
  • 10X focusable eyepiece
  • Coated antireflection glass optical components improve illumination and shadowline contrast
IFT40 Rhino Model
  • Sealed housing and eyepiece; IP67 rated water resistant and dustproof
  • Black polycarbonate housing reduces stray light
  • 0-30 arbitrary scale is divided into 0.5 increments; accuracy to ±0.25%
  • Eyepiece with large depth of focus
  • Built for production floor use; impact resistant to withstand repeated drops from three feet

Services

One year

Specifications

Specifications

0 to 30 Brix
Distilled Water
0.5%
Durable Plastic carrying case, sample application pipets, calibration adjustment wrench
1.33 to 1.38
±0.2
Ideal for applications including Metalworking coolants and lubricants, heat treating fluids, hydraulic fluids, firefighting Foam and plating baths
15°C to 38°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Reichert™ Industrial Fluid Testing Refractometers > RI range 1.3330 to 1.3860nD

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.