Learn More
Thermo Scientific™ Remel™ Agglutinating Sera, Haemophilus influenzae
Used for serological identification of H. influenzae by rapid slide agglutination test and CIE procedures.
Brand: Thermo Scientific™ R30166201
Description
Remel Agglutinating Sera are used for serological identification of H. influenzae by rapid slide agglutination test and CIE procedures.
Specifications
Agglutinating Sera, Haemophilus influenzae Type C | |
H. influenzae | |
Agglutinating Sera |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
6127643 | Certificate of Analysis | 03/01/2025 | R30166201 |
3800012 | Certificate of Analysis | 15/07/2024 | R30166201 |
3694177 | Certificate of Analysis | 05/01/2024 | R30166201 |
3644893 | Certificate of Analysis | 23/06/2023 | R30166201 |
3612058 | Certificate of Analysis | 21/03/2023 | R30166201 |