missing translation for 'onlineSavingsMsg'
Learn More
Rhodanine-3-acetic acid, 98%, Thermo Scientific Chemicals Product Code.: 11425896

Rhodanine-3-acetic acid, 98%, Thermo Scientific Chemicals

Product Code. 11425896
100 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11425896

Brand: Thermo Scientific Alfa Aesar B22244.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5718-83-2
Molecular Formula C5H5NO3S2
Molecular Weight (g/mol) 191.22
MDL Number MFCD00005491
InChI Key JGRMXPSUZIYDRR-UHFFFAOYSA-N
Synonym rhodanine-3-acetic acid, rhodanine-n-acetic acid, n-carboxymethylrhodanine, 3-rhodanineacetic acid, 3-thiazolidineacetic acid, 4-oxo-2-thioxo, 3-carboxymethyl rhodanine, n-carboxymethyl rhodanine, 2-4-oxo-2-thioxothiazolidin-3-yl acetic acid, 4-oxo-2-thioxo-3-thiazolidineacetic acid, 4-oxo-2-thioxo-3-thiazolidinylacetic acidShow MoreShow Less
PubChem CID 79793
IUPAC Name 2-(4-oxo-2-sulfanylidene-1,3-thiazolidin-3-yl)acetic acid
SMILES OC(=O)CN1C(=O)CSC1=S

Specifications

Melting Point 145°C to 148°C
Quantity 100 g
Beilstein 149513
Formula Weight 191.23
Percent Purity 98%
Chemical Name or Material Rhodanine-3-acetic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10248531 Certificate TypeCertificate of Analysis Date02/11/2023 Catalog NumberB22244.22,B22244.06,B22244.14
Lot Number10239910 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberB22244.22,B22244.06,B22244.14
Lot Number10162682 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB22244.22,B22244.06,B22244.14
Lot Number10115184 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB22244.22,B22244.06,B22244.14
Lot Number10179789 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB22244.22,B22244.06,B22244.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Rhodanine-3-acetic acid, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.