missing translation for 'onlineSavingsMsg'
Learn More

Pyrex™ Quickfit™ Rotary Evaporator Splash Head Adapter

Protects against foaming products that may seep into a condenser. Pyrex™ Quickfit™ Rotary Evaporator Splash Head Adapter have an anti-climb mechanism.

Brand:  Pyrex™ SH15/29

149.18 EUR valid until 2025-04-30
Use promo code "24155" to get your promotional price.


 View more versions of this product

Product Code. 12904075

  • 160.00 € / Each
Estimated Shipment: 30-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • For use with rotary evaporators
  • Protects against liquid ingress into the condenser 
  • Useful for liquids that have tendency to foam 
  • Splash heads feature a 29/32 socket
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Anti-climb
Rotary evaporators
Cone: 29/32, Socket: 29/32
Borosilicate Glass
1 Each
150 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Information

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Pyrex™ Quickfit™ Rotary Evaporator Splash Head Adapter > Cone Shape: 29/32; For use with: Useful for liquids that have tendency to Foam

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.