missing translation for 'onlineSavingsMsg'
Learn More

Gilac™ Round HACCP Food Contact Container, 75 L

Round HACCP Food Contact Container is food contact certified and designed for storage, food preparation and transporting foodstuffs.

Brand:  Gilac™ G612021

 View more versions of this product

Product Code. 17968981

  • 69.20 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • HACCP round container.
  • Food contact certified.
  • For storage, food preparation and transporting foodstuffs.
  • Highly durable.
  • Reinforced bottom with anti-wear skirt.
  • Reinforced handles to prevent distortion during carrying.
  • Optional, choice of two lids.
  • External Diameter includes handles.
  • Width corresponds to diameter without handles.
Specifications

Specifications

HACCP Food Container
19.8 gal.
670 mm
485 mm
19.8 gal.
21.3 in.
Polyethylene
White
19.1 in.
Round
75 L
540 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.