missing translation for 'onlineSavingsMsg'
Learn More
Bollé Safety Rush+ Platinum™ Smoke Safety Glasses Product Code.: 15206705
In Stock
to see stock.
Add to basket

Bollé Safety Rush+ Platinum™ Smoke Safety Glasses

Product Code. 15206705
This item is not returnable. View return policy

Product Code. 15206705

Brand: Bollé Safety RUSHPPSF

In Stock
to see stock.
Edge
Add to basket
Add to basket
This item is not returnable. View return policy

Ultra-sporty safety glasses with co-injected, ultra-flexible temples and anti-scratch/anti-fog lenses.

TRUSTED_SUSTAINABILITY

Specifications

Lens Coating Anti-fog, Scratch-resistant
Lens Tint Smoke
Product Type Rush RUSHPPSF Safety Spectacle
No. per Pack 1 Ea.
Weight (Metric) 25 g
  • Bollé Safety - Rush+
    Video Player is loading.
    Current Time 0:00
    Duration 1:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 1:00
     
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected

    Bolle Safety Rush+

  • Bollé Safety - Platinum coating
    Video Player is loading.
    Current Time 0:00
    Duration 1:06
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 1:06
     
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected

    Bolle Safety Platinum coating

  • Bollé Safety - Platinum efficiency demonstration
    Video Player is loading.
    Current Time 0:00
    Duration 3:09
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 3:09
     
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected

    Bolle Safety Platinum efficiency demonstration

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bollé Safety Rush+ Platinum™ Smoke Safety Glasses >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.