missing translation for 'onlineSavingsMsg'
Learn More

Ruthenium(III) bromide hydrate, Ru 25% min, Thermo Scientific Chemicals

Oxidation

Chemical Identifiers

CAS 314075-07-5
Molecular Formula Br3H2ORu
Molecular Weight (g/mol) 358.80
MDL Number MFCD00016314
InChI Key PMMMCGISKBNZES-UHFFFAOYSA-K
IUPAC Name tribromoruthenium hydrate
SMILES O.Br[Ru](Br)Br
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11337477
View Documents Promotion Details
Thermo Scientific Alfa Aesar
012628.03
1 g N/A
N/A
11347477
View Documents Promotion Details
Thermo Scientific Alfa Aesar
012628.06
5 g N/A
N/A
Description

Description

Ruthenium(III) bromide hydrate is used as a catalyst in chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

314075-07-5
358.80
PMMMCGISKBNZES-UHFFFAOYSA-K
O.Br[Ru](Br)Br
Br3H2ORu
MFCD00016314
tribromoruthenium hydrate

Specifications

314075-07-5
Br3H2ORu
UN3260
PMMMCGISKBNZES-UHFFFAOYSA-K
tribromoruthenium hydrate
340.78 (Anhydrous)
Characteristic
Ruthenium(III) bromide hydrate
Crystalline
MFCD00016314
Soluble in alcohol
O.Br[Ru](Br)Br
358.80
Ru 25% min.
Hygroscopic
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.

missing translation for 'einecsNumber' : 237-829-4

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only