missing translation for 'onlineSavingsMsg'
Learn More

(S)-verbenone, Thermo Scientific™

11.30 € - 58.00 €

Chemical Identifiers

CAS 1196-01-6
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.22
MDL Number MFCD00001343
InChI Key DCSCXTJOXBUFGB-JGVFFNPUSA-N
Synonym levoverbenone, --verbenone, l-verbenone, 1s---verbenone, --2-pinen-4-one, verbenone, unii-2xp0j7754u, 1s,5s-4,6,6-trimethylbicyclo 3.1.1 hept-3-en-2-one, verbenone, l, bicyclo 3.1.1 hept-3-en-2-one, 4,6,6-trimethyl-, 1s,5s
PubChem CID 92874
ChEBI CHEBI:78316
IUPAC Name (1S,5S)-2,6,6-trimethylbicyclo[3.1.1]hept-2-en-4-one
SMILES CC1=CC(=O)[C@H]2C[C@@H]1C2(C)C
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10199652
View Documents Promotion Details
Thermo Scientific Maybridge
SB01645DA
1 g
11.30 €
1g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10046434
View Documents Promotion Details
Thermo Scientific Maybridge
SB01645DE
5 g
32.20 €
5g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10537953
View Documents Promotion Details
Thermo Scientific Maybridge
SB01645EA
10 g
58.00 €
10g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

1196-01-6
150.22
DCSCXTJOXBUFGB-JGVFFNPUSA-N
92874
(1S,5S)-2,6,6-trimethylbicyclo[3.1.1]hept-2-en-4-one
C10H14O
MFCD00001343
levoverbenone, --verbenone, l-verbenone, 1s---verbenone, --2-pinen-4-one, verbenone, unii-2xp0j7754u, 1s,5s-4,6,6-trimethylbicyclo 3.1.1 hept-3-en-2-one, verbenone, l, bicyclo 3.1.1 hept-3-en-2-one, 4,6,6-trimethyl-, 1s,5s
CHEBI:78316
CC1=CC(=O)[C@H]2C[C@@H]1C2(C)C

Specifications

1196-01-6
97%
MFCD00001343
levoverbenone, --verbenone, l-verbenone, 1s---verbenone, --2-pinen-4-one, verbenone, unii-2xp0j7754u, 1s,5s-4,6,6-trimethylbicyclo 3.1.1 hept-3-en-2-one, verbenone, l, bicyclo 3.1.1 hept-3-en-2-one, 4,6,6-trimethyl-, 1s,5s
CC1=CC(=O)[C@H]2C[C@@H]1C2(C)C
150.22
CHEBI:78316
97%
0.9700g/mL
C10H14O
− 220.00 (20.00°C c=3−4,CHCL3)
DCSCXTJOXBUFGB-JGVFFNPUSA-N
(1S,5S)-2,6,6-trimethylbicyclo[3.1.1]hept-2-en-4-one
92874
150.22
(S)-Verbenone
Videos
SDS
Documents

Documents

Product Certifications