missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Asecos™ Safety Storage Cabinet V-MOVE-90
The asecos™ Safety Storage Cabinet V-MOVE-90 is robustly constructed and features automatic electronic opening and closing for added safety and convenience. Only suitable for indoor installation.
Brand: Asecos™ 3508604635102
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
- Cabinets for safe and approved storage of flammable hazardous substances in working areas
- Robust construction and longevity: robust, high-quality, scratch-resistant outer body with powder-coated surface
- Electronic opening and closing: the vertical drawers open and close at the push of a button
- Perfect access: the vertical drawers provide an overview and simple access to the stored containers
- Always closed: serial automatic door closing (TSA) after approximately 60 seconds with acoustic and visual signal before drawer closing
- No unauthorised use: vertical drawers lockable with cylinder locking (integration in an existing locking system possible) and locking state indicator (red/green)
- Easy alignment: adjusting aids on the base to compensate for uneven flooring
- Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system
- Fire resistance 90 minutes (EN 14470-1)
- CE compliant
Spezifikation
Type tested EN 14470-1, CE compliant. | |
Safety storage cabinet, V-Move-90™ | |
860x 819x 1966 mm | |
Safe and approved storage of hazardous substances | |
2 | |
Standard | |
Gray, Red | |
Type 90 safety cabinet |
2 red fronted tall drawers, each with 4 shelves and a bottom sump with perforated insert | |
775x 701x 1750 mm | |
Auto-close | |
V90.196.081. VDAC:0013 | |
8 | |
598 kg | |
Steel |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Asecos™ Safety Storage Cabinet V-MOVE-90 > Model: V90.196.081. VDAC:0013
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur