Learn More
Novacyt - Microgen Bioproducts Salmonella Latex Agglutination Test
Microgen Bioproducts Salmonella - A rapid, latex slide agglutination test for the presumptive identification of Salmonella in selective and/or enrichment cultures or isolates taken directly from selective agars. Simple one-step identification and confirmation of a range of pathogenic bacteria.
Brand: Novacyt - Microgen Bioproducts F42
Description
- A sensitive and highly specific test for the identification of Salmonella from clinical and food samples.
Specifications
ISO13485 ISO9001 | |
F42b Positive Control - 0.5 mL | |
Non-waived | |
3 Bottles (2.5 mL, 0.5 mL, 5.0 mL) | |
0.993 | |
A Latex Slide Agglutination Test for The Confirmatory Identification of Presumptive Salmonella Colonies From Selective Agar Plates | |
50 Tests | |
2 per test | |
Carton with Insert vial box | |
F42a Salmonella Latex Reagent - 2.5 mL | |
1 | |
4 minutes |
2°C to 8°C | |
No | |
Salmonella Antibodies | |
Bacterial Colonies | |
Agglutination Test | |
Bacteriological Loops, Pasteur Pipettes | |
Polythene Bottle | |
Monera | |
3 Bottles (2.5 mL, 0.5 mL, 5.0 mL) | |
Rabbit antiserum | |
Bacteria |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.