Learn More
Thermo Scientific™ Samco™ Special Purpose Transfer Pipettes
Eliminate plugging, packaging and autoclaving with these special purpose transfer pipettes, ideal for delivering drops onto slides or tiles.
25.80 € - 453.00 €
Product Code | Brand | Volume (Metric) | Sterility | Bulb Draw (Metric) | Description | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Sterility | Bulb Draw (Metric) | Description | Price | Quantity & Availability | |||
15377873
![]() |
Thermo Scientific™
292 |
1.0 mL | Non-sterile | 0.2 mL | Padl-Pet 25μL Drop Size |
27.30 €
Pack of 500 |
Estimated Shipment: 14-11-2025
Log in to see stock. |
|||
15337863
![]() |
Thermo Scientific™
273NL |
1.7 mL | Non-sterile | 0.9 mL | Samco 1.7 mL Transfer Pipets, Non-sterile, White Box |
26.60 €
Pack of 500 |
In Stock
Log in to see stock. |
|||
15788257
![]() |
Thermo Scientific™
273NL-250 |
1.7 mL | Non-sterile | 0.9 mL | Samco 1.7 mL Transfer Pipets, Non-sterile, 250/Bag, White Box |
453.00 €
Pack of 5000 |
Estimated Shipment: 12-08-2025
Log in to see stock. |
|||
11691829
![]() |
Thermo Scientific™
274NL |
4.8 mL | Non-sterile | 1.8 mL | Samco 4.8mL B/B-PET™ Transfer Pipets, Non-sterile, Graduated To 2mL, White Box |
25.80 €
Pack of 500 |
In Stock
Log in to see stock. |
|||
15377863
![]() |
Thermo Scientific™
274NL-1S |
4.8 mL | Sterile | 1.8 mL | Samco 4.8mL B/B-PET™ Transfer Pipets, Sterile, Graduated To 2mL, Individually Packed, White Box |
62.05 €
Pack of 500 |
Estimated Shipment: 25-09-2025
Log in to see stock. |
|||
15397863
![]() |
Thermo Scientific™
274NL-20S |
4.8 mL | Sterile | 1.8 mL | Samco 4.8 mL Transfer Pipets, Sterile, Graduated To 2 mL, 20/Bag, White Box |
38.30 €
Pack of 500 |
Estimated Shipment: 05-08-2025
Log in to see stock. |
|||
15387883
![]() |
Thermo Scientific™
336-10S |
5.0 mL | Sterile | 1.9 mL | Transfer pipet, 6 in., B/B-PET™, Standard Bulb, Sterile, 500 X 8 = 4000/CS |
47.05 €
Pack of 500 |
In Stock
Log in to see stock. ![]() |
|||
15317893
![]() |
Thermo Scientific™
336NL-1S |
5.0 mL | Sterile | 1.9 mL | Samco 5.0mL B/B-PET™ Transfer Pipets, 6 in., Sterile, Individually Packed, White Box |
65.05 €
Pack of 500 |
In Stock
Log in to see stock. ![]() |
|||
15327893
![]() |
Thermo Scientific™
336NL-20S |
5.0 mL | Sterile | 1.9 mL | Samco 5.0mL B/B-PET™ Transfer Pipets, 6 in., Sterile, 20/bag, White Box |
54.60 €
Pack of 500 |
Estimated Shipment: 22-08-2025
Log in to see stock. |
|||
15387873
![]() |
Thermo Scientific™
293 |
1.0 mL | Non-sterile | 0.2 mL | Padl-Pet 30μL Drop Size |
26.85 €
Pack of 500 |
In Stock
Log in to see stock. ![]() |
|||
15397873
![]() |
Thermo Scientific™
295 |
0.8 mL | Non-sterile | 0.2 mL | Padl-Pet 50μL Drop Size |
26.85 €
Pack of 500 |
Estimated Shipment: 18-09-2025
Log in to see stock. |
|||
Description
Eliminate plugging, packaging and autoclaving with Thermo Scientific™ Samco™ Special Purpose Transfer Pipettes, ideal for delivering drops onto slides or tiles. Single-unit design features a built-in pipettes bulb that eliminates the need for plugging, packaging, autoclaving; prevents cross-contamination.
- Shatterproof, nontoxic polyethylene
- Low-affinity surfaces –prevents proteins binding in research applications
- Can be frozen in liquid nitrogen, or heat sealed for storage
- Nonsterile
Specifications
LDPE |