Learn More
Sartorius Sterisart™ NF Sterility Testing System
BIOPHARMA
Provides completely closed system sterility testing of pharmaceuticals, open containers, closed containers, pre-filled syringes, powders, and medical devices. Sartorius Sterisart™ NF Sterility Testing System is optimal for use with isolators.
339.00 € - 840.00 €
Specifications
Capacity (English) 4 oz.
Capacity (Metric) 120 mL
For Use With (Application) Sterility Testing
Model Sterisart Consumable
Temperature (English) 122°F
View More Specs
Products 15
Product Code Brand Sample Composition Includes Sampling Option Sterilization Method Price Quantity & Availability  
Product Code Brand Sample Composition Includes Sampling Option Sterilization Method Price Quantity & Availability  
10687683
View Documents
Sartorius
16466--------ACD
Liquids Sterisart NF alpha with long dual needle metal spike, sterile-vented No Septum EtO This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11700965
BIOPHARMA
View Documents
Sartorius
16466--------GSD
Liquids Sterisart NF gamma with long dual-needle metal spike, sterile-vented and with partitioned connector for sterile sampling from Sterisart containers during incubation Septum Gamma irradiation
830.00 €
Pack of 10
In Stock
to see stock.
Edge
Add to basket
11790955
BIOPHARMA
View Documents
Sartorius
16466--------GBD
Liquids Sterisart NF gamma with long dual-needle metal spike, sterile vented No Septum Gamma irradiation
760.00 €
Pack of 10
Estimated Shipment: 10-07-2025
to see stock.
Add to basket
11720965
BIOPHARMA
View Documents
Sartorius
16467--------GBD
Liquids Sterisart NF gamma with long needle and protective plate, sterile venting needle No Septum Gamma irradiation
755.00 €
Pack of 10
Estimated Shipment: 16-06-2025
to see stock.
Add to basket
11740965
BIOPHARMA
View Documents
Sartorius
16468--------GBD
Liquids Sterisart NF gamma with luer or Luer Lock connector, long needle and sterile venting needle No Septum Gamma irradiation
755.00 €
Pack of 10
Estimated Shipment: 16-06-2025
to see stock.
Add to basket
11730975
BIOPHARMA
View Documents
Sartorius
16478--------GBD
Liquids Sterisart NF gamma with female Luer Lock connector No Septum Gamma irradiation
795.00 €
Pack of 10
Estimated Shipment: 10-07-2025
to see stock.
Add to basket
11750965
BIOPHARMA
View Documents
Sartorius
16469--------GBD
Liquids Sterisart NF gamma with adapter for pre-filled syringes and dual-needle metal spike, Syringes with and without needlessterile-vented No Septum Gamma irradiation
780.00 €
Pack of 10
Estimated Shipment: 16-06-2025
to see stock.
Add to basket
11770965
BIOPHARMA
View Documents
Sartorius
16470--------GBD
Liquids Sterisart NF gamma tubing system with two dual-needle metal spikes of different length No Septum Gamma irradiation
339.00 €
Pack of 10
Estimated Shipment: 25-09-2025
to see stock.
Add to basket
11720975
BIOPHARMA
View Documents
Sartorius
16477--------GBD
Liquids Sterisart NF gamma with long needle (side opening, with solid pointed tip, non-coring), protective plate, sterile venting needle No Septum Gamma irradiation
740.00 €
Pack of 10
Estimated Shipment: 29-04-2025
to see stock.
Add to basket
11790965
BIOPHARMA
View Documents
Sartorius
16475--------GBD
Easily dissolved powders Sterisart NF gamma with two dual-needle metal spikes of different length, one is sterile-vented No Septum Gamma irradiation
805.00 €
Pack of 10
Estimated Shipment: 10-07-2025
to see stock.
Add to basket
11710975
BIOPHARMA
View Documents
Sartorius
16476--------GBD
Liquids Sterisart NF gamma with short dual-needle metal spike, sterile-vented No Septum Gamma irradiation
755.00 €
Pack of 10
Estimated Shipment: 10-07-2025
to see stock.
Add to basket
11730965
BIOPHARMA
View Documents
Sartorius
16467--------GSD
Liquids Long needle and protective plate, sterile venting needle and with partitioned connector for sterile sampling from Sterisart containers during incubation Septum Gamma irradiation
820.00 €
Pack of 10
Estimated Shipment: 27-05-2025
to see stock.
Add to basket
15972530
BIOPHARMA
View Documents
Sartorius
16476--------GSD
Liquids Sterisart NF gamma Septum with short dual-needle metal spike, sterile vented and with partitioned connector for sterile sampling from containers during incubation Septum Gamma irradiation
805.00 €
Pack of 10
Estimated Shipment: 16-06-2025
to see stock.
Add to basket
11760965
BIOPHARMA
View Documents
Sartorius
16469--------GSD
Liquids Sterisart NF gamma Septum with adapter for prefi lled syringes and dualneedle metal spike, sterile-vented and with a partitioned connector for sterile sampling from Sterisart containers during incubation Septum Gamma irradiation
835.00 €
Pack of 10
Estimated Shipment: 10-07-2025
to see stock.
Add to basket
11700975
BIOPHARMA
View Documents
Sartorius
16475--------GSD
Easily dissolved powders Sterisart NF gamma Septum with two dual-needle metal spikes of different length, one is sterile-vented, and with a partitioned connector for sterile sampling from Sterisart® containers during incubation Septum Gamma irradiation
840.00 €
Pack of 10
Estimated Shipment: 10-07-2025
to see stock.
Add to basket
Description
Description
International pharmacopoeias require the complete sterility of pharmaceutical products that are injected into the blood stream or that otherwise enter the body below the skin surface. As a manufacturer of such products, you are required to supply proof of sterility of the final product batch. Sterisart™ NF is a completely closed system for the sterility testing of pharmaceutical products. It is based on the membrane filter method, however it eliminates the procedure of manipulating the filters. By this the main risk of a secondary contamination and false positive results is eliminated. A peristaltic pump transfers the sample into the filtration units, and after rinsing, the filtration units are filled with media and used for incubation of the filters without any contact to the environment.
  • reliable Sartochem™ membrane
  • high retention of microbes
  • low adsorption
  • high mechanical stability
  • pre-installed color-coded tube clamps
  • easy-to-read graduated marks
  • user-friendly (several practical adapters available)
  • product-/lot number identification
  • gas-impermeable packaging for protection against sterilants
Specifications
Specifications
4 oz.
Sterility Testing
122°F
15.7 cm
3 bar at 20°C
120 mL
Sterisart Consumable
50°C
0.45 μm
Sterile
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Sartorius Sterisart™ NF Sterility Testing System