missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Scintillation Vials

Designed with heavy walls to minimize solvent losses. Kartell™ Scintillation Vials are compatible with all liquid scintillation counters.

Brand:  Kartell™ 0093400

 View more versions of this product

Product Code. 10172611

  • 191.00 € / Pack of 2000
Estimated Shipment: 09-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Unbreakable
  • Available in macro or mini sizes
  • Supplied with screw caps
  • CE marked, IVD approved 
Specifications

Specifications

Translucent
Polyethylene
Polypropylene
52.5 mm
13.71 mm
4 mL
Screw Cap
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Kartell™ Scintillation Vials > Capacity: 4mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.