missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ SecureFit™ 400X Safety Glasses, Scotchgard™ Anti-Fog / Anti-Scratch Coating (K&N)
Click to view available options
Frame Color:
Black
Blue, Gray
Green, Black
Green, Clear
Yellow, Black
Lens Tint:
Amber
Clear
Gray
Light Gray, Indoor/Outdoor
Orange, Mirror
Vermillion
Description
- Safety glasses with 3M™ Pressure Diffusion Temple Technology for comfort
- Wraparound design to improve the security of fit and prevent slipping
- Padded temple touchpoints for increased comfort over the ears
- Soft nose bridge for a more comfortable fit
- Contoured brow guard provides additional coverage from flying particles
- Superior anti-fog and anti-scratch properties meets the K&N marking requirements of EN166
- Frame marking: SF400 Series, 3M EN166 FT

Specifications
Specifications
Certifications/Compliance | CE Approved |
Lens Coating | Anti-fog |
Lens Tint | Clear |
Product Type | Protective Eyewear |
Frame Color | Green, Clear |
Temple Style | Pressure Diffusion Temple Technology |
Lens Material | Polycarbonate |
Frame Material | Plastic |
UV Protection | UV400 |
Product Suggestions
Customers who viewed this item also viewed.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
3M™ SecureFit™ 400X Safety Glasses, Scotchgard™ Anti-Fog / Anti-Scratch Coating (K&N) >
Spot an opportunity for improvement?Share a Content Correction