missing translation for 'onlineSavingsMsg'
Learn More

Selenium standard solution, for AAS, 1mg/mL Se in 10% HCl, Thermo Scientific Chemicals

Brand:  Thermo Scientific Acros 196281000

23.20 EUR valid until 2025-05-31
Use promo code "24127" to get your promotional price.



 View more versions of this product

Product Code. 10695832

  • 24.42 € / 100mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for atomic adsorption spectroscopy
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Selenium standard solution
7782-49-2, 7647-01-0, 7732-18-5
100 mL
Se
Solution
995 to 1005 μg/mL of Se in 10% HCl (20°C)
[Se]
Soluble in water
6326970
78.96
1 mg/ml Se in 10% HCl
Plastic bottle
4.81
MFCD00134090 MFCD00011224
14, 8431
BUGBHKTXTAQXES-UHFFFAOYSA-N
selenium
78.97
CHEBI:27568
AAS
Product Suggestions

Product Suggestions

Videos
Safety and Handling
Show More
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0449906 Certificate TypeCertificate of Analysis Date31/07/2024 Catalog Number196285000,196281000
Lot NumberA0461046 Certificate TypeCertificate of Analysis Date14/03/2024 Catalog Number196285000,196281000
Lot NumberA0460493 Certificate TypeCertificate of Analysis Date26/02/2024 Catalog Number196285000,196281000
Lot NumberA0410402 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number196285000,196281000
Lot NumberA0430032 Certificate TypeCertificate of Analysis Date11/11/2022 Catalog Number196285000,196281000
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Selenium standard solution, for AAS, 1mg/mL Se in 10% HCl, Thermo Scientific Chemicals > 100mL; Plastic bottle

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

RUO – Research Use Only