Learn More
BrandTech™ VACUUBRAND™ Service Kits for Oil-Free Diaphragm Vacuum Pumps
Original VACUUBRAND replacement parts
Brand: BrandTech™ VACUUBRAND™ 20696860
Description
- Includes appropriate diaphragms, valves, and sealing rings, if appropriate
- Does not include spanner wrench
Specifications
Model ME4 NT, MZ2 NT, MZ2 NT Vario Oil free diaphragm pump | |
Service Kit |
Product Suggestions
Customers who viewed this item also viewed