Learn More
SGE™ Gastight Syringes: Fixed Needle Models
Suitable for liquid and gas samples
58.95 € - 307.00 €
Specifications
Accuracy ±1%
For Use With (Application) Suitable for gases or liquids for maximum inertness
Includes Plunger protection, needle glued into barrel with epoxy solvent-resistant glue
Product Type Gastight Syringe
Needle Point Style Fixed Bevel
View More Specs
Products 10
Product Code Brand Volume (Metric) Needle Gauge Model Price Quantity & Availability  
Product Code Brand Volume (Metric) Needle Gauge Model Price Quantity & Availability  
10229250
View Documents
SGE™
002200
10 μL 26 G 10F-GT
58.95 €
Each
15298443
View Documents
SGE™
002202
10 μL 26 G 10F-GT
307.00 €
Pack of 6
15208453
View Documents
SGE™
002208
10 μL 26 G 10F-GT-7
65.30 €
Each
10167450
View Documents
SGE™
004200
50 μL 25 G 25F-GT
65.60 €
Each
10035540
View Documents
SGE™
005200
100 μL 25 G 25F-GT
73.20 €
Each
10702772
View Documents
SGE™
006200
250 μL 25 G 250F-GT
74.55 €
Each
10106930
View Documents
SGE™
007200
500 μL 25 G 500F-GT
73.20 €
Each
10605093
View Documents
SGE™
008102
1 mL 22 G 1MDF-GT
75.85 €
Each
12780710
View Documents
SGE™
008502
2.5 mL 22 G 2.5MDF-GT
77.10 €
Each
10546772
View Documents
SGE™
003200
25 μL 25 G 25F-GT
66.65 €
Each
Description
Description
Suitable for liquid and gas samples
  • 10μL to 100mL capacities
  • Constructed from borosilicate glass, with PTFE plunger tips and Kel-F™ Luer cones (where applicable)
  • Needles align with zero mark on barrel to reduce dead volume, improve accuracy and precision to <±1%
  • Replaceable plungers on all models
  • Fixed-needle models have needle glued into barrel with epoxy solvent-resistant glue
Specifications
Specifications
±1%
Plunger protection, needle glued into barrel with epoxy solvent-resistant glue
Fixed Bevel
Suitable for gases or liquids for maximum inertness
Gastight Syringe
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
SGE™ Gastight Syringes: Fixed Needle Models