Learn More
Woodsafe™ Sharpsbin and Hazardous waste containers with yellow double lid

Woodsafe is a cost-effective solution for reducing the use of fossil based plastic by 80% and CO2 emissions by 66%, documented in in a full LCA. Produced with residue from the forest industry using green energy. Available in sizes from 0.5 L to 50 L. Single and double lid versions.
Brand: Woodsafe™ 1060222
266.05 EUR valid until 2025-05-30
Use promo code "24004" to get your promotional price.
Alert:
Follow instructions for safe and correct use
Description
- Woodsafe™ offer a sustainable clinical waste management solution produced with residue from the Swedish forest industry
- Lowers the use of Fossil-based plastic by 80%!
- Lowers the carbon footprint by more than 60%, proved in full LCA
- Made from 100% Biocomposite
- The lid can be securely permanently closed for transportation to the final disposal
- My Green Labs ACT Label
- Follows same process as yellow bins and can be used to transport the same substances
- Natural color and smell from wood - No migration of substances
- Compliant with UN3291
- Tested according to ISO 23907-1:2019
- UN ADR Approved
- Biocomposite have the same properties as fossil-based plastic and can be used exactly the same way
- NOTE: String between lids is for delivery purpose, remove/break before use!
- Stop burning plastic
Specifications
6 L | |
Temporary and permanent closure | |
Round | |
1.32 gal. | |
31.5 cm X 18.6 cm X 15.5 cm (12.4 x 7.32 x 6.1 in.) | |
30 | |
Yes | |
Autoclavable | |
Safe Disposal of sharps and potentially Infectious Waste or medical waste | |
Biocomposite |
EN ISO 23907-1:2019. UN3291, ADR Approved | |
Yellow | |
Environmental friendly Sharpsbin | |
Brown, Yellow | |
Single-use | |
Round | |
Non-sterile | |
Bin, Lid, and Label | |
Cardboard | |
Biohazard Waste Container (Benchtop) |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Lower climate impact and ACT label