missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Silicone Rubber Stopper
Description
- Non-toxic, translucent
- Temperature range -60°C to 200°C
- High electrical resistance
- High resistance to many chemicals including ozone
Specifications
1/Pk. | |
Translucent | |
96 mm | |
Silicone Stopper |
−60°C to 200°C | |
86 mm | |
Silicone | |
86 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ Silicone Rubber Stopper >
Spot an opportunity for improvement?Share a Content Correction