Learn More
Silver nanopowder, APS 20-40nm, 99.9% (metals basis), Thermo Scientific Chemicals
Description
Silver nanopowder is involved in the coating of silver and is used for medicinal purposes and in medical devices. Its larger surface area to volume ratio and greater activity of nano-silver are utilized in modern biochemical applications and drug delivery. It is also used in the fabrication of graphite epoxy nanocomposites and optical fiber sensors. Furthermore, it is involved in the formulation of water based ink.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
UN Number | UN3077 |
Quantity | 5 g |
Solubility Information | Soluble in nitric acid. |
Percent Purity | 99.9% |
Odor | Characteristic |
Particle Size | APS 20 to 40nm |
Form | Nanopowder |
Assay | (metals basis) |
Chemical Name or Material | Silver nanopowder, No Oxide layer |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
Y30H001 | Certificate of Analysis | 12/04/2022 | 045509.14,045509.06 |
N09I038 | Certificate of Analysis | 12/04/2022 | 045509.14,045509.06 |
E06X026 | Certificate of Analysis | 08/12/2021 | 045509.14,045509.06 |
X27B004 | Certificate of Analysis | 07/12/2021 | 045509.14,045509.06 |
U03E073 | Certificate of Analysis | 07/12/2021 | 045509.14,045509.06 |
Safety and Handling


- Silver nanopowder
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Reproductive toxicity Category 1A
- H360-May damage fertility or the unborn child.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P403-Store in a well-ventilated place.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : Silver, nanoform
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.