missing translation for 'onlineSavingsMsg'
Learn More
Silver slug, 3.175mm (0.125in) dia x 3.175mm (0.125in) length, 99.99% (metals basis), Thermo Scientific Chemicals Product Code.: 11320188

Silver slug, 3.175mm (0.125in) dia x 3.175mm (0.125in) length, 99.99% (metals basis), Thermo Scientific Chemicals

Product Code. 11320188
5 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11320188

Brand: Thermo Scientific Alfa Aesar 042924.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Silver slug is used in thermal evaporation and electrodes (Schottky diode and fast diode).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-22-4
Molecular Formula Ag
Molecular Weight (g/mol) 107.87
MDL Number MFCD00003397
InChI Key BQCADISMDOOEFD-UHFFFAOYSA-N
Synonym argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
PubChem CID 23954
ChEBI CHEBI:9141
IUPAC Name silver
SMILES [Ag]

Specifications

Quantity 5 g
Solubility Information Insoluble in water.
Percent Purity 99.99%
Odor Characteristic
Weight ≈0.26g/slug
Form Slug
Assay (metals basis)
Chemical Name or Material Silver slug, 3.175mm (0.125 in.) dia. x 3.175mm (0.125 in.) length

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberL28W037 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number042924.06,042924.14
Lot NumberM17E024 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number042924.06,042924.14
Lot NumberQ03F025 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number042924.06,042924.14
Lot NumberH25X054 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number042924.06,042924.14
Lot NumberP11G037 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number042924.06,042924.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Silver slug, 3.175mm (0.125in) dia x 3.175mm (0.125in) length, 99.99% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.