missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Slide-A-Lyzer™ G3 Dialysis Cassettes, 3.5K MWCO
GREENER_CHOICE

Thermo Scientific Slide-A-Lyzer™ G3 Dialysis Cassettes facilitate the rapid and trouble-free dialysis of sample volumes from 0.5 to 125 mL. Unlike standard flat tubing, these innovative devices do not require knots or clips that can lead to leaking and sample loss.

Brand:  Thermo Scientific™ A52970

205.70 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.


 View more versions of this product

Product Code. 17884010

  • 248.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

Buffer Exchange, Protein
Cassette
3.5 kDa
6 cassettes
Cellulose Membrane, Dialysis
125 mL
Slide-A-Lyzer™
Less waste
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberFP2939407 Certificate TypeCertificate of Analysis Date16/12/2024 Catalog NumberA52970
Lot NumberFP2868364 Certificate TypeCertificate of Analysis Date12/07/2024 Catalog NumberA52970
Lot NumberFP286855 Certificate TypeCertificate of Analysis Date04/07/2024 Catalog NumberA52970
Lot NumberFP2846222 Certificate TypeCertificate of Analysis Date16/02/2024 Catalog NumberA52970
Lot NumberFP2830071 Certificate TypeCertificate of Analysis Date16/02/2024 Catalog NumberA52970
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Slide-A-Lyzer™ G3 Dialysis Cassettes, 3.5K MWCO >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.