Learn More
Ibidi μ-Slide VI 0.4 μ-Pattern RGD, sqr20, pit110, hex
μ-Slide VI 0.4 μ-Pattern RGD, sqr20, pit110, hex is a 6 channel slide with micropatterned surface.
Brand:  Ibidi 83601
Encompass
Product Code. 17286233
  • POA / Pack of 10
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
Description
Description
  • A 6 channel slide with micropatterned surface
  • bottom material: #1.5 Polymer Coverslip with passivated surface (biologically inert) and RGD pattern
  • sterilized
Specifications
Specifications
Microscope Slide
10
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Ibidi μ-Slide VI 0.4 μ-Pattern RGD, sqr20, pit110, hex