missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Blue Disposable Coveralls, 4500 Series
Use for non-hazardous applications. The Blue Disposable Coveralls, 4500 Series offers Category I protection from polypropylene fabric.
52.90 € - 60.90 €
Specifications
Product Type | General Purpose Coveralls |
---|---|
Gender | Unisex |
Material | Polyethylene |
Color | Blue |
Disposable | Single-use |
Product Code | Brand | Size | Height (English) | Height (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Height (English) | Height (Metric) | Price | Quantity & Availability | ||||||
13493676
|
3M™
7000089601 |
2X-Large | 73 to 76 in. | 186 to 194 cm | N/A | N/A | N/A | |||||
13403686
|
3M™
7000089599 |
Large | 69 to 71 in. | 174 to 181 cm | N/A | N/A | N/A | |||||
13483676
|
3M™
7000089598 |
Medium | 66 to 69 in. | 167 to 176 cm |
52.90 €
Pack of 20 |
Estimated Shipment: 14-04-2025
Log in to see stock. |
||||||
13413686
|
3M™
7000089600 |
X-Large | 70 to 74 in. | 179 to 187 cm |
52.90 €
Pack of 20 |
Estimated Shipment: 08-05-2025
Log in to see stock. |
||||||
13473676
|
3M™
7000089597 |
Small | 64 to 67 in. | 164 to 170 cm | N/A | N/A | N/A | |||||
13423686
|
3M™
7000089602 |
3X-Large | 76 to 78 in. | 194 to 200 cm | N/A | N/A | N/A | |||||
13433686
|
3M™
7000089603 |
4X-Large | 78 to 81 in. | 200 to 206 cm | N/A | N/A | N/A | |||||
Description
CE Simple, Category 1 coverall is designed for domestic DIY use, light-duty industrial cleaning and maintenance applications, or environments where hygiene and cleanliness are important.- Excellent economic solution to keep the wearer clean and dry
- Suitable for non-hazardous applications
- Breathable and light-duty protection
- Strong, durable 45gsm polypropylene fabric
- Storm flap over zip
- Elasticated hood, cuffs, ankles, and waist
Specifications
General Purpose Coveralls | |
Polyethylene | |
Single-use | |
Elastic | |
2-panel | |
Serged | |
Category I | |
Light Industrial Cleaning, Maintenance |
Unisex | |
Blue | |
Elastic | |
Elastic | |
Front Zipper with Storm Flap | |
Non-sterile | |
4500B |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
3M™ Blue Disposable Coveralls, 4500 Series