Learn More
Fisherbrand™ Soda Lime Glass Rimless Test Tube
Store test reagents and material. Fisherbrand™ soda lime glass rimless test tubes come in different sizes to meet your needs.
Brand:  Fisherbrand™ FB63114
 View more versions of this product
Product Code. 11912198
Description
Description
  • Soda Lime Glass
  • Rimless
  • Disposable 
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ISO 4142
Soda Lime Glass
No
150 mm
100 Pack
16 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Soda Lime Glass Rimless Test Tube > Dimensions: 16 dia. x 150mmH