missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Soda Lime Glass Rimless Test Tube
Store test reagents and material. Fisherbrand™ soda lime glass rimless test tubes come in different sizes to meet your needs.
Brand: Fisherbrand™ FB63115
This item is not returnable.
View return policy
Description
- Soda Lime Glass
- Rimless
- Disposable

Specifications
ISO 4142 | |
Soda Lime Glass | |
No |
150 mm | |
100 Pack | |
18 mm |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 12
Fisherbrand Focus on glassware and plasticware
File size: 66.4 MB
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ Soda Lime Glass Rimless Test Tube > Dimensions: 18 dia. x 150mmH
Spot an opportunity for improvement?Share a Content Correction