missing translation for 'onlineSavingsMsg'
Learn More

Sodium Azide, Certified AR for Analysis, Fisher Chemical™

94.00 € - 204.00 €

Chemical Identifiers

CAS 26628-22-8
Molecular Formula N3Na
Molecular Weight (g/mol) 65.011
MDL Number 3536
InChI Key PXIPVTKHYLBLMZ-UHFFFAOYSA-N
Synonym sodium azide, azide, sodium, natriumazid, nemazyd, smite, kazoe, azoture de sodium, sodium azide na n3, hydrazoic acid, sodium salt, natriumazid german
PubChem CID 33557
ChEBI CHEBI:278547
IUPAC Name sodium;azide
SMILES [N-]=[N+]=[N-].[Na+]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10050520
SDS View Documents
Fisher Chemical
S/2380/48
100 g
94.00 €
100g
Please to purchase this item. Need a web account? Register with us today!
10274060
SDS View Documents
Fisher Chemical
S/2380/53
500 g
204.00 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

26628-22-8
65.011
PXIPVTKHYLBLMZ-UHFFFAOYSA-N
33557
sodium;azide
N3Na
3536
sodium azide, azide, sodium, natriumazid, nemazyd, smite, kazoe, azoture de sodium, sodium azide na n3, hydrazoic acid, sodium salt, natriumazid german
CHEBI:278547
[N-]=[N+]=[N-].[Na+]

Specifications

300°C
26628-22-8
10
N3Na
sodium azide, azide, sodium, natriumazid, nemazyd, smite, kazoe, azoture de sodium, sodium azide na n3, hydrazoic acid, sodium salt, natriumazid german
[N-]=[N+]=[N-].[Na+]
65.011
CHEBI:278547
Plastic powder jar
275°C
White
Powder/Solid
3536
PXIPVTKHYLBLMZ-UHFFFAOYSA-N
sodium;azide
33557
65.01g/mol
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Sodium azide

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 1
  • Specific target organ toxicity after repeated exposure Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1

Hazard Statement
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H373-May cause damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
  • EUH032-Contact with acids liberates very toxic gas.

Precautionary Statement
  • P330-Rinse mouth.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P350-IF ON SKIN: Gently wash with plenty of soap and water.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.