missing translation for 'onlineSavingsMsg'
Learn More

Sodium Chloride, Certified AR for Analysis, meets analytical specification of Ph.Eur, Fisher Chemical™

ClNa, CAS Number-7647-14-5, saline, table salt, salt, sodium chloride, common salt, rock salt, dendritis, halite, sodium chloride nacl, purex, 1kg, 1461 deg.C, CHEBI:26710, White, 58.44g/mol, FAPWRFPIFSIZLT-UHFFFAOYSA-M, sodium;chloride, 801 deg.C, 3477, Plastic powder jar, 5.0 to 8.0, Solid, 5234

Brand:  Fisher Chemical S/3160/60

34.65 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 10428420

  • 45.75 € / 1kg

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

ClNa, CAS Number-7647-14-5, saline, table salt, salt, sodium chloride, common salt, rock salt, dendritis, halite, sodium chloride nacl, purex, 1kg, 1461 deg.C, CHEBI:26710, White, 58.44g/mol, FAPWRFPIFSIZLT-UHFFFAOYSA-M, sodium;chloride, 801 deg.C, 3477, Plastic powder jar, 5.0 to 8.0, Solid, 5234

TRUSTED_SUSTAINABILITY
Specifications

Specifications

1461°C
7647-14-5
5.0 to 8.0
1 kg
ClNa
sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
[Na+].[Cl-]
58.44
CHEBI:26710
Plastic powder jar
801°C
White
Solid
99.5%
MFCD00003477
FAPWRFPIFSIZLT-UHFFFAOYSA-M
sodium chloride
5234
58.44g/mol
1mmHg 865
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium Chloride, Certified AR for Analysis, meets analytical specification of Ph.Eur, Fisher Chemical™ > 1kg, Plastic powder jar

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.