missing translation for 'onlineSavingsMsg'
Learn More
Learn More

- Home
- Products
- Chemicals
- Salts and Inorganics
- Inorganic Salts
- Alkali Metal Salts
- Sodium chloride, meets analytical specification of Ph.Eur
Favorite item to keep tabs on it!
Saved!
Go to My Lists
You have reached the limit of 350 favorited items.
Manage Your Favorites
Sodium chloride, meets analytical specification of Ph.Eur
Product Code. 119840511 kg, Plastic powder jar
Click to view available options
Quantity:
1 kg
3 kg
10 kg
25 kg
Packaging:
Plastic keg
Plastic powder jar
This item is not returnable.
View return policy
Product Code. 11984051
Brand: Fisher Chemical S/3161/60
37.50 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Explore more special offers
Use promo code "25339" to get your promotional price.
Explore more special offers
This item is not returnable.
View return policy
Specifications
Chemical Identifiers
CAS | 7647-14-5 |
Molecular Formula | ClNa |
Molecular Weight (g/mol) | 58.44 |
MDL Number | MFCD00003477 |
InChI Key | FAPWRFPIFSIZLT-UHFFFAOYSA-M |
Synonym | sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex |
PubChem CID | 5234 |
ChEBI | CHEBI:26710 |
SMILES | [Na+].[Cl-] |
Specifications
Quantity | 1 kg |
IUPAC Name | sodium;chloride |
Packaging | Plastic powder jar |
Product Suggestions
You may be interested in these alternatives
-
Product Code. 11904061
-
-
Customers who viewed this item also viewed. This information does not imply a recommendation or representation of any kind and any action taken upon the information provided is strictly at your own risk. The products shown have not been qualified by Fisher Scientific for any particular purpose, application or use and it is the responsibility of the purchaser to ensure that the performance of the product is suitable for the purchasers specific use or intended application.
Viewing 1-4 of
-
Catalog No. 10092740
27,80 € / 1kg -
Catalog No. 10428420
45,75 € / 1kg -
Catalog No. 10735921
181,00 € / 5kg -
Catalog No. 10112640
33,95 € / 1kg -
Catalog No. 10055850
30,55 € / 500g -
Catalog No. 10316380
67,75 € / 2.50 L -
Catalog No. 10162180
124,00 € / 2.50 L -
Catalog No. 10675692
49,35 € / 1kg -
Catalog No. 10304980
160,00 € / 2.50 L -
Catalog No. 10703811
382,00 € / 500g -
Catalog No. 10793611
154,00 € / 1kg -
Catalog No. 10117113
34,57 € / 100g -
Catalog No. 10401841
27,10 € / 500mL -
Catalog No. 15681810
37,34 € / 25g
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sodium chloride, meets analytical specification of Ph.Eur >
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.
Spot an opportunity for improvement?Share a Content Correction